Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026435.hg.1 |
TC05001912.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026435.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026435.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026435.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026435.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026436.hg.1 |
TC05001912.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026436.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026436.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026436.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026436.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026438.hg.1 |
TC05001912.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026438.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026438.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186328.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186328.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186328.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186328.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186328.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186328.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186328.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186328.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186328.hg.1 |
TC0500012394.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500186327.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186327.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186327.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186327.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186327.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186327.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186327.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186327.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186327.hg.1 |
TC0500012394.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186326.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186326.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186326.hg.1 |
TC0500012394.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500186331.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186331.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186331.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186331.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186331.hg.1 |
TC0500012394.hg.1 |
Clariom D TC |