| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014262.hg.1 |
ENST00000245663.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014262.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014262.hg.1 |
ENST00000395104.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014262.hg.1 |
ENST00000480766.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014262.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014262.hg.1 |
TC20001039.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000460757.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000460757.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000480766.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
ENST00000480766.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014263.hg.1 |
ENST00000245663.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014263.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014263.hg.1 |
ENST00000395104.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014263.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014263.hg.1 |
TC20001039.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014266.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014266.hg.1 |
TC20001039.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014260.hg.1 |
ENST00000245663.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014260.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014260.hg.1 |
ENST00000395104.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014260.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20014260.hg.1 |
TC20001039.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000460757.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000460757.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000480766.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
ENST00000480766.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009833.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168703.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168703.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168703.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168703.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168703.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168703.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168703.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168703.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168703.hg.1 |
TC2000009833.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000168707.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168707.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168707.hg.1 |
TC2000009833.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000168706.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168706.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168706.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168706.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168706.hg.1 |
TC2000009833.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000168701.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168701.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168701.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168701.hg.1 |
ENST00000480766.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168701.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168701.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168701.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168701.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168701.hg.1 |
ENST00000480766.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168701.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168701.hg.1 |
TC2000009833.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000168705.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168705.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168705.hg.1 |
TC2000009833.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000168696.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168696.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168696.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168696.hg.1 |
ENST00000460757.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168696.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168696.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168696.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168696.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168696.hg.1 |
ENST00000460757.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168696.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168696.hg.1 |
TC2000009833.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000168702.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168702.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168702.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168702.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168702.hg.1 |
ENST00000245663.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168702.hg.1 |
ENST00000302995.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168702.hg.1 |
ENST00000395104.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168702.hg.1 |
NM_025224.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168702.hg.1 |
TC2000009833.hg.1 |
Clariom D TC |