Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01010229_s1
          See other MRGPRG GE Assays ›
          Gene Symbol:
          MRGPRG
          Gene Name:
          MAS related GPR family member G
          Gene Aliases:
          GPR169, MRGG
          Chromosome Location:
          Chr.11: 3217944 - 3218813 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01010229_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MRGPRG
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164377.1 NP_001157849.1 1 66 69
          GenBank mRNA AY255583.1 - 1 66 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100009836.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001292.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3303176 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 15286 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P10974173 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3303176 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3303176 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606797 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606749 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606789 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004085 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606754 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606776 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606736 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004098 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606778 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606788 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606785 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606793 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606772 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606791 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606759 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606763 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606752 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606745 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606779 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606739 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606782 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606747 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606781 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606784 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004123 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606777 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004101 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606761 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606783 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606741 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606795 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606798 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606734 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004083 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004144 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004107 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606764 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004093 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606741 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004107 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606788 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606763 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606772 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606793 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606752 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606784 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606754 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004144 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606797 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606739 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606776 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004123 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606779 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606785 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606747 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606795 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606783 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606789 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606778 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004098 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606745 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004093 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606734 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606736 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606761 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606782 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606764 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606759 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004083 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606749 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606777 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606781 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004101 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606791 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28004085 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07606798 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7945858 - -
          Affymetrix Human Gene 1.1 ST Array 7945857 - -
          Affymetrix Human Transcriptome Array 2.0 PSR11016466.hg.1 ENST00000332314.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016466.hg.1 NM_001164377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016466.hg.1 TC11001292.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100009836.hg.1 ENST00000332314.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009836.hg.1 ENST00000332314.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009836.hg.1 NM_001164377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009836.hg.1 NM_001164377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000420873.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000420873.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000420873.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000420873.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000420873.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000420873.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000434798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000434798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000434798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000434798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000434798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000434798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000531711.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000531711.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000531711.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000531711.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000541883.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 ENST00000541883.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 NR_027138.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 NR_027138.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 NR_027138.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 NR_027138.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 NR_027138.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006611.hg.1 NR_027138.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100174387.hg.1 ENST00000332314.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174387.hg.1 NM_001164377.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100174387.hg.1 ENST00000332314.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174387.hg.1 NM_001164377.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100174387.hg.1 TC1100009836.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000420873.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000420873.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000420873.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000420873.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000420873.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000420873.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000434798.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000434798.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000434798.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000434798.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000434798.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000434798.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000531711.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000531711.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000531711.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000531711.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000541883.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 ENST00000541883.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 NR_027138.1 Refseq
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 NR_027138.1 Refseq
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 NR_027138.1 Refseq
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 NR_027138.1 Refseq
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 NR_027138.1 Refseq
          Affymetrix Human Clariom D Assay TC1100006611.hg.1 NR_027138.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009836.hg.1 ENST00000332314.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009836.hg.1 ENST00000332314.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009836.hg.1 NM_001164377.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009836.hg.1 NM_001164377.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100174385.hg.1 ENST00000332314.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174385.hg.1 NM_001164377.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100174385.hg.1 ENST00000332314.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174385.hg.1 NM_001164377.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100174385.hg.1 TC1100009836.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A044784 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A044784 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 661290 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3299421 NM_001164377.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline