| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15006503.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15006503.hg.1 |
ENST00000406419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15006503.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15006503.hg.1 |
TC15000728.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000406419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000406419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000406419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000406419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000406419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000560753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000560753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000560753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
ENST00000560753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000406419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000406419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000406419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000406419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000406419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000560753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000560753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000560753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
ENST00000560753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007972.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500157186.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500157186.hg.1 |
ENST00000406419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500157186.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500157186.hg.1 |
ENST00000299529.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500157186.hg.1 |
ENST00000406419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500157186.hg.1 |
NM_004378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500157186.hg.1 |
TC1500007972.hg.1 |
Clariom D TC |