| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006182.hg.1 |
ENST00000227520.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006182.hg.1 |
ENST00000339492.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006182.hg.1 |
ENST00000545580.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006182.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006182.hg.1 |
TC11000518.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000545580.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000545580.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000545580.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000545580.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
ENST00000545580.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000545580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000545580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000545580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000545580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
ENST00000545580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007781.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100155008.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100155008.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100155008.hg.1 |
ENST00000545580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100155008.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100155008.hg.1 |
ENST00000227520.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100155008.hg.1 |
ENST00000535217.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100155008.hg.1 |
ENST00000545580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100155008.hg.1 |
NM_024098.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100155008.hg.1 |
TC1100007781.hg.1 |
Clariom D TC |