Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01011446_m1
          See other CBL GE Assays ›
          Gene Symbol:
          CBL
          Gene Name:
          Cbl proto-oncogene
          Gene Aliases:
          C-CBL, CBL2, FRA11B, NSLL, RNF55
          Chromosome Location:
          Chr.11: 119206276 - 119308149 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01011446_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CBL
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005188.3 NP_005179.2 5 - 6 1013 72
          XM_011543057.2 XP_011541359.1 5 - 6 1013 72
          XM_017018511.1 XP_016874000.1 4 - 5 448 72
          GenBank mRNA BC132733.1 - 5 - 6 942 40146755 72
          BC136463.1 - 5 - 6 942 9020452 72
          X57110.1 - 5 - 6 1019 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100009245.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001089.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CBL

          Panther Classification:

          Molecular Function -

          ligase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          cell surface receptor signaling pathway
          epidermal growth factor receptor signaling pathway
          transforming growth factor beta receptor signaling pathway
          fibroblast growth factor receptor signaling pathway
          positive regulation of phosphatidylinositol 3-kinase signaling
          protein ubiquitination
          negative regulation of epidermal growth factor receptor signaling pathway
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic process
          positive regulation of epidermal growth factor receptor signaling pathway
          positive regulation of receptor-mediated endocytosis
          cell-cell adhesion
          phosphotyrosine binding
          transcription factor activity, sequence-specific DNA binding
          ubiquitin-protein transferase activity
          signal transducer activity
          calcium ion binding
          protein binding
          zinc ion binding
          ligase activity
          SH3 domain binding
          protein kinase binding
          ephrin receptor binding
          ubiquitin protein ligase activity
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11013860.hg.1 ENST00000264033.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11013860.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11013860.hg.1 TC11001089.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11013859.hg.1 ENST00000264033.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11013859.hg.1 NM_005188.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11013859.hg.1 TC11001089.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009245.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100169338.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100169338.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100169338.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100169338.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100169338.hg.1 TC1100009245.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100169339.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100169339.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100169339.hg.1 ENST00000264033.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100169339.hg.1 NM_005188.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100169339.hg.1 TC1100009245.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline