| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036390.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036390.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036390.hg.1 |
TC02002263.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036389.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036389.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036389.hg.1 |
TC02002263.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200203051.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203051.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203051.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203051.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203051.hg.1 |
TC0200014085.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
ENST00000464621.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014085.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203050.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203050.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203050.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203050.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203050.hg.1 |
TC0200014085.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200203052.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203052.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203052.hg.1 |
ENST00000263707.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203052.hg.1 |
NM_014553.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203052.hg.1 |
TC0200014085.hg.1 |
Clariom D TC |