| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031603.hg.1 |
TC01002088.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031603.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031603.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031609.hg.1 |
TC01002088.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031609.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031609.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031606.hg.1 |
TC01002088.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031606.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031606.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031607.hg.1 |
TC01002088.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031607.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031607.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000462849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000462849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000462849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000462849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000462849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000462849.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000478606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000478606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000478606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000478606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000478606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031605.hg.1 |
TC01002088.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031605.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01031605.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191555.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191555.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191555.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191555.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191555.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191555.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191555.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191555.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191555.hg.1 |
TC0100012456.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100191559.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191559.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191559.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191559.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191559.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191559.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191559.hg.1 |
TC0100012456.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100191562.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191562.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191562.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191562.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191562.hg.1 |
TC0100012456.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100191558.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191558.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191558.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191558.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191558.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191558.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191558.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191558.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191558.hg.1 |
TC0100012456.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100191561.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191561.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191561.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191561.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191561.hg.1 |
TC0100012456.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000462849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000462849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000462849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000462849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000462849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000462849.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000478606.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000478606.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000478606.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000478606.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000478606.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012456.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191557.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191557.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191557.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191557.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191557.hg.1 |
ENST00000330388.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191557.hg.1 |
ENST00000468365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191557.hg.1 |
ENST00000486627.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100191557.hg.1 |
NM_001014980.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100191557.hg.1 |
TC0100012456.hg.1 |
Clariom D TC |