| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000591606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000591606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000591606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000591606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000591606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000592769.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000592769.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000592769.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000592769.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000592769.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000585392.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000585392.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000585392.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000585392.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000585392.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000587579.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000587579.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000587579.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588599.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588599.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588599.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588599.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588599.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588599.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000588599.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000590212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000590212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000590212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000590212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000590212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000590212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
ENST00000590212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000342669.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000391932.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000585392.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000585392.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000585392.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000585392.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000585392.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000587367.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000587579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000587579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000587579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000588599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000590212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000590212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000590212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000590212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000590212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000590212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
ENST00000590212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_004597.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010951.hg.1 |
NM_177542.3 |
Refseq |