Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01019254_m1
          See other LOC650226 GE Assays ›
          Gene Symbol:
          LOC650226
          Gene Name:
          ankyrin repeat domain 26 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 56423704 - 56448375 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01019254_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC650226
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029420.1 1 - 2 151 96
          GenBank mRNA DB043272.1 - 1 - 2 151 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700011180.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001417.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Oligo Microarray 4x44K 23276 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P59592277 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P59592277 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05506246 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731173 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05506250 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731193 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731187 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05506252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731184 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731169 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05506247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05506244 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15946300 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731188 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05506242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15946293 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15946288 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731185 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731175 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15946297 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05506252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731193 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05506247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731187 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05506250 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731169 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731185 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15946300 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15946297 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05506246 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731175 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05506244 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731184 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731188 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05506242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731173 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15946288 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15946293 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25731174 - -
          Agilent Human 1B Microarray G4111A A_32_P722725 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07021912.hg.1 TC07001417.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07021912.hg.1 NR_029420.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011150.hg.1 ENST00000432235.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011150.hg.1 ENST00000432235.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011150.hg.1 ENST00000432235.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011150.hg.1 ENST00000432235.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011150.hg.1 ENST00000432235.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011150.hg.1 ENST00000432235.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011150.hg.1 ENST00000432235.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07021913.hg.1 TC07001417.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07021913.hg.1 NR_029420.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700007279.hg.1 ENST00000413879.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007279.hg.1 ENST00000413879.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007279.hg.1 ENST00000413879.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007279.hg.1 ENST00000413879.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007279.hg.1 ENST00000413879.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007279.hg.1 ENST00000413879.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07021917.hg.1 TC07001417.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07021917.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07021917.hg.1 NR_029420.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07021915.hg.1 TC07001417.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07021915.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07021915.hg.1 NR_029420.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07021916.hg.1 TC07001417.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07021916.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07021916.hg.1 NR_029420.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07021918.hg.1 TC07001417.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07021918.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07021918.hg.1 NR_029420.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 ENST00000430165.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 ENST00000430165.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 NR_029420.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 NR_029420.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 NR_029420.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 NR_029420.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 NR_029420.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 NR_029420.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011180.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay TC0700007279.hg.1 ENST00000413879.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007279.hg.1 ENST00000413879.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007279.hg.1 ENST00000413879.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007279.hg.1 ENST00000413879.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007279.hg.1 ENST00000413879.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007279.hg.1 ENST00000413879.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011150.hg.1 ENST00000432235.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011150.hg.1 ENST00000432235.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011150.hg.1 ENST00000432235.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011150.hg.1 ENST00000432235.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011150.hg.1 ENST00000432235.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011150.hg.1 ENST00000432235.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011150.hg.1 ENST00000432235.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179893.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179893.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179893.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700179893.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179893.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179893.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700179893.hg.1 TC0700011180.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700179891.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179891.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179891.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700179891.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179891.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179891.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700179891.hg.1 TC0700011180.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700179887.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179887.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700179887.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179887.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700179887.hg.1 TC0700011180.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700179889.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179889.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700179889.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179889.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700179889.hg.1 TC0700011180.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700179888.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179888.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700179888.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179888.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700179888.hg.1 TC0700011180.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700179892.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179892.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700179892.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179892.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700179892.hg.1 TC0700011180.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700179896.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179896.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700179896.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700179896.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700179896.hg.1 TC0700011180.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 ENST00000422212.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 ENST00000430165.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 ENST00000430165.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 ENST00000443198.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 NR_029420.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011180.hg.1 NR_029420.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A032845 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A032846 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A032845 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A032846 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 467496 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline