Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01022023_m1
          See other GABPA GE Assays ›
          Gene Symbol:
          GABPA
          Gene Name:
          GA binding protein transcription factor alpha subunit
          Gene Aliases:
          E4TF1-60, E4TF1A, NFT2, NRF2, NRF2A, RCH04A07
          Chromosome Location:
          Chr.21: 25734947 - 25772460 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01022023_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GABPA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001197297.1 NP_001184226.1 9 - 10 1281 118
          NM_002040.3 NP_002031.2 9 - 10 1718 118
          XM_005260938.4 XP_005260995.1 9 - 10 1353 118
          XM_011529520.2 XP_011527822.1 10 - 11 1757 118
          XM_011529521.2 XP_011527823.1 11 - 12 1848 118
          XM_017028313.1 XP_016883802.1 9 - 10 1364 118
          GenBank mRNA AK223164.1 - 9 - 10 1320 118
          AK313545.1 - 9 - 10 1515 118
          BC035031.2 - 9 - 10 1351 5264778 118
          BX647755.1 - 9 - 10 1659 118
          D13318.1 - 9 - 10 1323 118
          EU159453.1 - 8 - 9 1140 118
          JQ313900.1 - 8 - 9 1131 118
          U13044.1 - 9 - 10 1316 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100006787.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000078.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GABPA

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          transcription from RNA polymerase II promoter
          mitochondrion organization
          cell differentiation
          negative regulation of megakaryocyte differentiation
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          protein binding
          transcription regulatory region DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000487266.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000487266.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000487266.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000487266.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000487266.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 ENST00000487266.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000431.hg.1 ENST00000354828.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000431.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000431.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000431.hg.1 NM_002040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000431.hg.1 TC21000078.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2100145802.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145802.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145802.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145802.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100145802.hg.1 TC2100006787.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100145802.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145802.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145802.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145802.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100145803.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145803.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145803.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145803.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100145803.hg.1 TC2100006787.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100145803.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145803.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2100145803.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100145803.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000354828.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000400075.3 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000487266.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000487266.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000487266.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000487266.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000487266.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 ENST00000487266.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_001197297.1 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006787.hg.1 NM_002040.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 167077 - -
          ABI Human Genome Survey Microarray Version 2.0 226728 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline