| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009572.hg.1 |
ENST00000439973.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009572.hg.1 |
ENST00000439973.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009572.hg.1 |
ENST00000439973.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009572.hg.1 |
ENST00000441907.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009572.hg.1 |
ENST00000441907.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009572.hg.1 |
ENST00000441907.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009572.hg.1 |
ENST00000441907.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009572.hg.1 |
ENST00000414107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009572.hg.1 |
ENST00000439973.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009572.hg.1 |
ENST00000439973.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009572.hg.1 |
ENST00000439973.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009572.hg.1 |
ENST00000441907.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009572.hg.1 |
ENST00000441907.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009572.hg.1 |
ENST00000441907.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009572.hg.1 |
ENST00000441907.1 |
Ensembl |