| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05014344.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05014344.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05014344.hg.1 |
TC05000997.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000503175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000503175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000507151.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000507151.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000507151.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000507151.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000507151.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000514533.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000514533.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
ENST00000514533.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05014345.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05014345.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05014345.hg.1 |
TC05000997.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500167059.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500167059.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500167059.hg.1 |
TC0500009585.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500167060.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500167060.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500167060.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500167060.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500167060.hg.1 |
TC0500009585.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500167061.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500167061.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500167061.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500167061.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500167061.hg.1 |
TC0500009585.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000503175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000503175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000507151.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000507151.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000507151.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000507151.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000507151.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000514533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000514533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
ENST00000514533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009585.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500167062.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500167062.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500167062.hg.1 |
ENST00000310389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500167062.hg.1 |
NM_173664.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500167062.hg.1 |
TC0500009585.hg.1 |
Clariom D TC |