Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01025917_m1
          See other CBR3 GE Assays ›
          Gene Symbol:
          CBR3
          Gene Name:
          carbonyl reductase 3
          Gene Aliases:
          HEL-S-25, SDR21C2, hCBR3
          Chromosome Location:
          Chr.21: 36134901 - 36146562 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01025917_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CBR3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001236.3 NP_001227.1 1 - 2 515 127
          XM_011529772.2 XP_011528074.1 1 - 2 579 127
          GenBank mRNA AB004854.1 - 1 - 2 287 127
          AB041012.1 - 1 - 2 568 127
          AB124847.1 - 1 - 2 462 127
          AK223429.1 - 1 - 2 393 127
          BC002812.2 - 1 - 2 375 3637434 127
          CR541709.1 - 1 - 2 127
          FJ224313.1 - 1 - 2 287 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100007037.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000152.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CBR3

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          phylloquinone catabolic process
          cognition
          oxidation-reduction process
          3-keto sterol reductase activity
          carbonyl reductase (NADPH) activity
          NADPH binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR21001249.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001249.hg.1 NM_001236.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21001249.hg.1 TC21000152.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21001248.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001248.hg.1 NM_001236.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21001248.hg.1 TC21000152.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2100007037.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007037.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007037.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007037.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007037.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007037.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007037.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007037.hg.1 NM_001236.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007037.hg.1 NM_001236.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007037.hg.1 NM_001236.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007037.hg.1 NM_001236.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007037.hg.1 NM_001236.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007037.hg.1 NM_001236.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21001247.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001247.hg.1 NM_001236.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21001247.hg.1 TC21000152.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC2100007037.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007037.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007037.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007037.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007037.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007037.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007037.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007037.hg.1 NM_001236.3 Refseq
          Affymetrix Human Clariom D Assay TC2100007037.hg.1 NM_001236.3 Refseq
          Affymetrix Human Clariom D Assay TC2100007037.hg.1 NM_001236.3 Refseq
          Affymetrix Human Clariom D Assay TC2100007037.hg.1 NM_001236.3 Refseq
          Affymetrix Human Clariom D Assay TC2100007037.hg.1 NM_001236.3 Refseq
          Affymetrix Human Clariom D Assay TC2100007037.hg.1 NM_001236.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100147485.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100147485.hg.1 NM_001236.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100147485.hg.1 TC2100007037.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100147485.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100147485.hg.1 NM_001236.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100147486.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100147486.hg.1 NM_001236.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100147486.hg.1 TC2100007037.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100147486.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100147486.hg.1 NM_001236.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100147489.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100147489.hg.1 NM_001236.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100147489.hg.1 TC2100007037.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100147489.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100147489.hg.1 NM_001236.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100147487.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100147487.hg.1 NM_001236.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100147487.hg.1 TC2100007037.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100147487.hg.1 ENST00000290354.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100147487.hg.1 NM_001236.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 145553 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline