| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007495.hg.1 |
ENST00000430110.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007495.hg.1 |
ENST00000438774.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007495.hg.1 |
ENST00000451188.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007495.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007495.hg.1 |
TC06004087.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007497.hg.1 |
ENST00000451188.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007497.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007497.hg.1 |
TC06004087.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007494.hg.1 |
ENST00000430110.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007494.hg.1 |
ENST00000438774.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007494.hg.1 |
ENST00000451188.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007494.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007494.hg.1 |
TC06004087.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014133.hg.1 |
ENST00000438774.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014133.hg.1 |
ENST00000438774.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014133.hg.1 |
ENST00000438774.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014133.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014133.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014133.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014133.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014133.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014133.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014133.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014133.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014133.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007496.hg.1 |
ENST00000430110.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007496.hg.1 |
ENST00000438774.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007496.hg.1 |
ENST00000451188.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007496.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007496.hg.1 |
TC06004087.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600198803.hg.1 |
ENST00000438774.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198803.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198803.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600198803.hg.1 |
ENST00000438774.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198803.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198803.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600198803.hg.1 |
TC0600014133.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600198805.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198805.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600198805.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198805.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600198805.hg.1 |
TC0600014133.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600014133.hg.1 |
ENST00000438774.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014133.hg.1 |
ENST00000438774.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014133.hg.1 |
ENST00000438774.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014133.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014133.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014133.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014133.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014133.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014133.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014133.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014133.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014133.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600198804.hg.1 |
ENST00000438774.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198804.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198804.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600198804.hg.1 |
ENST00000438774.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198804.hg.1 |
ENST00000451188.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600198804.hg.1 |
NM_001137560.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600198804.hg.1 |
TC0600014133.hg.1 |
Clariom D TC |