Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060371.hg.1 |
TC01006388.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060371.hg.1 |
ENST00000366606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060371.hg.1 |
ENST00000400947.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060371.hg.1 |
ENST00000408888.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060371.hg.1 |
NM_001161533.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060371.hg.1 |
NM_015014.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01060371.hg.1 |
NR_027762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
ENST00000408888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
ENST00000429912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
ENST00000447801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
ENST00000468751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
ENST00000474086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
ENST00000476261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
NM_001161533.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
NM_015014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
NR_027762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
ENST00000408888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
ENST00000429912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
ENST00000447801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
ENST00000468751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
ENST00000474086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
ENST00000476261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
NM_001161533.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
NM_015014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
NR_027762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244684.hg.1 |
TC0100018567.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
ENST00000408888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
ENST00000429912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
ENST00000447801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
ENST00000468751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
ENST00000474086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
ENST00000475960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
ENST00000476261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
NM_001161533.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
NM_015014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
NR_027762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
ENST00000408888.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
ENST00000429912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
ENST00000447801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
ENST00000468751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
ENST00000474086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
ENST00000475960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
ENST00000476261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
NM_001161533.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
NM_015014.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
NR_027762.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244685.hg.1 |
TC0100018567.hg.1 |
Clariom D TC |