Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01026531_g1
          See other TAS1R3 GE Assays ›
          Gene Symbol:
          TAS1R3
          Gene Name:
          taste 1 receptor member 3
          Gene Aliases:
          T1R3
          Chromosome Location:
          Chr.1: 1331324 - 1335320 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01026531_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TAS1R3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152228.2 NP_689414.1 5 - 6 1602 58
          GenBank mRNA AK122959.1 - 2 - 3 140 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100006502.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000035.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 G-protein coupled receptor Ion Channel 3' Most Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of sweet taste
          G-protein coupled receptor signaling pathway
          sensory perception of sweet taste
          sensory perception of umami taste
          G-protein coupled receptor activity
          taste receptor activity
          sweet taste receptor activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3359084 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3286973 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3260989 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3085 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3084 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 47598 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 10191 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 18592 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 6048 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3260989 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3260989 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15008235 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001791 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001792 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00001787 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_152228_at - -
          Affymetrix Human Gene 1.0 ST Array 7896923 - -
          Affymetrix Human Gene 1.0 ST Array 7896926 - -
          Affymetrix Human Gene 1.0 ST Array 7896925 - -
          Affymetrix Human Gene 1.0 ST Array 7896927 - -
          Affymetrix Human Gene 1.0 ST Array 7896924 - -
          Affymetrix Human Gene 1.0 ST Array 7896928 - -
          Affymetrix Human Gene 1.0 ST Array 7896922 - -
          Affymetrix Human Gene 1.1 ST Array 7896921 - -
          Affymetrix Human Genome U219 Array 11740945_PM_at - -
          Affymetrix Human Genome U219 Array 11740945_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR01000440.hg.1 TC01000035.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01000440.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01000440.hg.1 NM_152228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006502.hg.1 NM_152228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006502.hg.1 NM_152228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006502.hg.1 NM_152228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006502.hg.1 NM_152228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006502.hg.1 NM_152228.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006502.hg.1 NM_152228.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100006502.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100006502.hg.1 NM_152228.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006502.hg.1 NM_152228.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006502.hg.1 NM_152228.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006502.hg.1 NM_152228.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006502.hg.1 NM_152228.2 Refseq
          Affymetrix Human Clariom D Assay TC0100006502.hg.1 NM_152228.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100144772.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100144772.hg.1 NM_152228.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100144772.hg.1 ENST00000339381.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100144772.hg.1 NM_152228.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100144772.hg.1 TC0100006502.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014300 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014300 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 138471 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2108135 NM_152228.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1798875 AK122959.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2108135 NM_152228.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2108135 NM_152228.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2108135 NM_152228.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2108135 NM_152228.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1798875 AK122959.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2108135 NM_152228.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2108135 NM_152228.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2108135 NM_152228.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-xmktn:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0