Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01026908_m1
          See other ST8SIA3 GE Assays ›
          Gene Symbol:
          ST8SIA3
          Gene Name:
          ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3
          Gene Aliases:
          SIAT8C, ST8SiaIII
          Chromosome Location:
          Chr.18: 57352557 - 57368931 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01026908_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ST8SIA3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015879.3 NP_056963.2 1 - 2 471 71
          GenBank mRNA AB208996.1 - 1 - 2 542 71
          AF003092.2 - 1 - 2 253 71
          AF004668.2 - 1 - 2 2204 71
          AK289517.1 - 1 - 2 316 71
          AK313047.1 - 1 - 2 264 71
          BC074909.2 - 1 - 2 229 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1800007394.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC18000194.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ST8SIA3

          Gene Ontology Categories:

          Function(s) Process(es)

          ganglioside biosynthetic process
          protein glycosylation
          N-glycan processing
          glycosphingolipid biosynthetic process
          glycoprotein metabolic process
          oligosaccharide metabolic process
          sialylation
          alpha-N-acetylneuraminate alpha-2,8-sialyltransferase activity
          sialyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups
          sialic acid binding
          identical protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000586360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000586360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000586360.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000590600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 ENST00000590600.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18002295.hg.1 ENST00000324000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18002295.hg.1 ENST00000541833.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18002295.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18002295.hg.1 TC18000194.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR18002294.hg.1 ENST00000324000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18002294.hg.1 ENST00000541833.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18002294.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18002294.hg.1 TC18000194.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR18002292.hg.1 ENST00000324000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18002292.hg.1 ENST00000541833.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18002292.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18002292.hg.1 TC18000194.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR18002296.hg.1 ENST00000324000.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18002296.hg.1 ENST00000541833.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18002296.hg.1 NM_015879.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18002296.hg.1 TC18000194.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1800150703.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1800150703.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800150703.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1800150703.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800150703.hg.1 TC1800007394.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800150705.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1800150705.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800150705.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1800150705.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800150705.hg.1 TC1800007394.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800150702.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1800150702.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800150702.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1800150702.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800150702.hg.1 TC1800007394.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000586360.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000586360.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000586360.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000590600.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 ENST00000590600.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay TC1800007394.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800150704.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1800150704.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800150704.hg.1 ENST00000324000.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1800150704.hg.1 NM_015879.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800150704.hg.1 TC1800007394.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline