Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01027517_m1
          See other KATNAL2 GE Assays ›
          Gene Symbol:
          KATNAL2
          Gene Name:
          katanin catalytic subunit A1 like 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 46917594 - 47102243 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01027517_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KATNAL2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001353899.1 NP_001340828.1 14 - 15 1668 61
          NM_001353900.1 NP_001340829.1 14 - 15 1665 61
          NM_001353901.1 NP_001340830.1 14 - 15 1939 61
          NM_001353902.1 NP_001340831.1 13 - 14 1569 61
          NM_001353903.1 NP_001340832.1 11 - 12 1387 61
          NM_001353904.1 NP_001340833.1 10 - 11 1559 61
          NM_001353905.1 NP_001340834.1 14 - 15 1680 61
          NM_001353906.1 NP_001340835.1 11 - 12 1387 61
          NM_001353907.1 NP_001340836.1 11 - 12 1658 61
          NM_001353908.1 NP_001340837.1 10 - 11 1288 61
          NM_001353909.1 NP_001340838.1 10 - 11 1559 61
          NM_001367621.1 NP_001354550.1 12 - 13 1135 61
          NM_001387690.1 NP_001374619.1 14 - 15 1931 61
          NM_031303.3 NP_112593.2 11 - 12 1102 61
          NR_148563.1 13 - 14 1566 61
          XM_005258361.4 XP_005258418.1 10 - 11 877 61
          XM_011526219.4 XP_011524521.1 14 - 15 1668 61
          XM_011526220.3 XP_011524522.1 14 - 15 1668 61
          XM_011526221.4 XP_011524523.1 14 - 15 1668 61
          XM_011526223.4 XP_011524525.1 14 - 15 1668 61
          XM_017026031.2 XP_016881520.1 10 - 11 877 61
          XM_047437877.1 XP_047293833.1 14 - 15 1665 61
          XM_047437878.1 XP_047293834.1 14 - 15 1665 61
          XM_047437879.1 XP_047293835.1 14 - 15 1668 61
          XM_047437880.1 XP_047293836.1 14 - 15 1665 61
          GenBank mRNA AK309134.1 - 4 - 5 711 61
          AL512748.1 - 1 - 2 104 61
          BC034999.2 - 11 - 12 1075 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1800007233.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC18000169.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KATNAL2

          Gene Ontology Categories:

          Function(s) Process(es)

          meiotic spindle organization
          cytoplasmic microtubule organization
          microtubule severing
          nucleotide binding
          protein binding
          ATP binding
          microtubule binding
          microtubule-severing ATPase activity
          isomerase activity
          ATPase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1800008611.hg.1 ENST00000592747.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008611.hg.1 ENST00000592747.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000245121.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000356157.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585383.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585383.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585383.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585383.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585469.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585469.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585469.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585469.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585469.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585469.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585469.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585469.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585469.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585469.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585469.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000585469.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000586198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000586198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000586198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000586198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000586198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000588433.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000588433.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000588433.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000588433.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000588433.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000588433.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000588433.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000588433.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000588433.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000588433.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000591522.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000591522.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000591522.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000591522.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000592005.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000592005.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000592005.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000592005.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000592005.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000592005.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 ENST00000592005.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007233.hg.1 NM_031303.3 Refseq
          Affymetrix Human Clariom D Assay TC1800008611.hg.1 ENST00000592747.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800008611.hg.1 ENST00000592747.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline