| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01033840.hg.1 |
ENST00000376957.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01033840.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01033840.hg.1 |
TC01002195.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000465788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000465788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000465788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000465788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000465788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000475189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000475189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000475189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000475189.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000487300.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000487300.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000487300.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000487300.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000487300.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000490101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000490101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000490101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
ENST00000490101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000459997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000465788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000465788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000465788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000465788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000465788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000475189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000475189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000475189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000475189.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000487300.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000487300.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000487300.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000487300.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000487300.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000490101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000490101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000490101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
ENST00000490101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012869.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195023.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195023.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195023.hg.1 |
ENST00000376957.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100195023.hg.1 |
NM_003132.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100195023.hg.1 |
TC0100012869.hg.1 |
Clariom D TC |