Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01027931_s1
          See other HIST1H2AE GE Assays ›
          Gene Symbol:
          HIST1H2AE
          Gene Name:
          histone cluster 1, H2ae
          Gene Aliases:
          H2A.1, H2A.2, H2A/a, H2AFA
          Chromosome Location:
          Chr.6: 26216920 - 26217483 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01027931_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HIST1H2AE
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021052.2 NP_066390.1 1 488 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600007285.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000173.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HIST1H2AE

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin silencing
          negative regulation of cell proliferation
          DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04625285 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24813556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04625286 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 20682 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 14745 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P59045 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P59045 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 17493 - -
          Agilent Human Genome CGH Microarray 44B A_14_P115530 - -
          Agilent Human Genome CGH Microarray 44A A_14_P115530 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 633 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P59045 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P59045 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04625287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24813556 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04625285 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04625286 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24813556 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04625287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04625286 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04625285 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P59045 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P115530 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04625286 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04625287 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04625286 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04625287 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04625284 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04625285 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P115530 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P115530 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P59045 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 4967 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 72310 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P115530 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625318 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625346 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625344 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625339 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625286 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625281 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625309 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625324 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625313 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625326 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625304 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625336 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625315 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625322 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625329 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625331 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04625342 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P115530 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_021052_at - -
          Affymetrix Human Gene 1.0 ST Array 8117410 - -
          Affymetrix Human Gene 1.0 ST Array 8117413 - -
          Affymetrix Human Gene 1.0 ST Array 8117414 - -
          Affymetrix Human Gene 1.0 ST Array 8117412 - -
          Affymetrix Human Gene 1.0 ST Array 8117409 - -
          Affymetrix Human Gene 1.0 ST Array 8117411 - -
          Affymetrix Human Gene 1.1 ST Array 8117408 - -
          Affymetrix HT HG-U133+ PM Array Plate 214469_PM_at - -
          Affymetrix Human Genome U219 Array 11731694_s_at - -
          Affymetrix Human Genome U219 Array 11731694_PM_s_at - -
          Affymetrix Human Genome U219 Array 11731693_at - -
          Affymetrix Human Genome U219 Array 11731693_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR06001758.hg.1 TC06000173.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06001758.hg.1 ENST00000303910.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06001758.hg.1 NM_021052.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06001757.hg.1 TC06000173.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06001757.hg.1 ENST00000303910.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06001757.hg.1 NM_021052.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600011137.hg.1 ENST00000630628.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600149363.hg.1 NM_021052.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600149363.hg.1 NM_021052.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600149363.hg.1 TC0600007285.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600149364.hg.1 NM_021052.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600149364.hg.1 NM_021052.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600149364.hg.1 TC0600007285.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600011137.hg.1 ENST00000630628.1 Ensembl
          Human Genome U133 Plus 2.0 Array 214469_at - -
          Human Genome U133 Plus 2.0 Array 214469_PM_at - -
          Human Genome U133A 2.0 Array 214469_at - -
          Human Genome U133A 2.0 Array 214469_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_021052_at - -
          AffymetrixPathwork Diagnostics Pathchip pw512757 - -
          Human Genome U95 Set 35127_PM_at - -
          Human Genome U95 Set 35127_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214469_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214469_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214469_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214469_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214469_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214469_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214469_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214469_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214469_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214469_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214469_at7 - -
          Human Genome U133 Set 214469_at - -
          Human Genome U133 Set 214469_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A031111 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A031111 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 119320 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1756849 NM_021052.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1756849 BC093836.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1756849 BC093862.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1756849 NM_021052.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1756849 BC093836.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1756849 BC093862.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1756849 NM_021052.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1756849 BC093836.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1756849 BC093862.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1756849 NM_021052.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1756849 BC093836.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1756849 BC093862.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1756849 NM_021052.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1756849 BC093836.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1756849 BC093862.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_19557649-S NM_021052.2 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_1822-S - -
          Illumina human-6 v1.0 expression beadchip 0006020338 - -
          Illumina humanRef-8 v1.0 expression beadchip 0006020338 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1756849 NM_021052.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1756849 BC093836.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1756849 BC093862.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1756849 NM_021052.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1756849 BC093836.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1756849 BC093862.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 5860400 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_1822 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1756849 NM_021052.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1756849 BC093836.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1756849 BC093862.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1756849 NM_021052.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1756849 BC093836.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1756849 BC093862.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1756849 NM_021052.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1756849 BC093836.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1756849 BC093862.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1756849 NM_021052.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1756849 BC093836.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1756849 BC093862.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1756849 NM_021052.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1756849 BC093836.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1756849 BC093862.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline