| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013234.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013234.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013234.hg.1 |
TC14001160.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000479171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000556755.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000556755.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000556755.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
ENST00000556755.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013232.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013232.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013232.hg.1 |
TC14001160.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013233.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013233.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013233.hg.1 |
TC14001160.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000479171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000556755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000556755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000556755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
ENST00000556755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009258.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400168378.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168378.hg.1 |
ENST00000556755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168378.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400168378.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168378.hg.1 |
ENST00000556755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168378.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400168378.hg.1 |
TC1400009258.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400168374.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168374.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400168374.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168374.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400168374.hg.1 |
TC1400009258.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400168380.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168380.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168380.hg.1 |
TC1400009258.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400168376.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168376.hg.1 |
ENST00000556755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168376.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400168376.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168376.hg.1 |
ENST00000556755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168376.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400168376.hg.1 |
TC1400009258.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400168379.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168379.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400168379.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168379.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400168379.hg.1 |
TC1400009258.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400168375.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168375.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400168375.hg.1 |
ENST00000247219.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400168375.hg.1 |
NM_199047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400168375.hg.1 |
TC1400009258.hg.1 |
Clariom D TC |