| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007451.hg.1 |
ENST00000568264.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007451.hg.1 |
ENST00000568264.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16002819.hg.1 |
ENST00000287594.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16002819.hg.1 |
ENST00000396385.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16002819.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16002819.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16002819.hg.1 |
TC16000189.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16002817.hg.1 |
ENST00000396385.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16002817.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16002817.hg.1 |
TC16000189.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1600007451.hg.1 |
ENST00000568264.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007451.hg.1 |
ENST00000568264.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011356.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189539.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189539.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189539.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189539.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189539.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189539.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189539.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189539.hg.1 |
NM_173803.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189539.hg.1 |
TC1600011356.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600189537.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189537.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189537.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189537.hg.1 |
ENST00000396385.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189537.hg.1 |
ENST00000564148.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189537.hg.1 |
NM_001128423.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189537.hg.1 |
TC1600011356.hg.1 |
Clariom D TC |