Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01033302_m1
          See other CD200 GE Assays ›
          Gene Symbol:
          CD200
          Gene Name:
          CD200 molecule
          Gene Aliases:
          MOX1, MOX2, MRC, OX-2
          Chromosome Location:
          Chr.3: 112332224 - 112362812 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01033302_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CD200
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004196.3 NP_001004196.2 5 - 6 1040 70
          NM_001318826.1 NP_001305755.1 4 - 5 960 70
          NM_001318828.1 NP_001305757.1 3 - 4 883 70
          NM_001318830.1 NP_001305759.1 3 - 4 878 70
          NM_005944.6 NP_005935.4 4 - 5 965 70
          XM_005247481.4 XP_005247538.1 5 - 6 1214 70
          XM_005247482.2 XP_005247539.1 4 - 5 844 70
          XM_005247485.2 XP_005247542.1 4 - 5 1155 70
          XM_005247486.2 XP_005247543.1 3 - 4 652 70
          XM_005247488.2 XP_005247545.1 3 - 4 757 70
          XM_017006452.1 XP_016861941.1 5 - 6 1541 70
          XM_017006453.1 XP_016861942.1 3 - 4 807 70
          XM_017006454.1 XP_016861943.1 5 - 6 1530 70
          XM_017006455.1 XP_016861944.1 4 - 5 1165 70
          XM_017006456.1 XP_016861945.1 5 - 6 891 70
          XM_017006457.1 XP_016861946.1 4 - 5 839 70
          XM_017006458.1 XP_016861947.1 5 - 6 1531 70
          XM_017006459.1 XP_016861948.1 4 - 5 1150 70
          GenBank mRNA AF063591.1 - 4 - 5 756 70
          AK074972.1 - 4 - 5 730 70
          AK129797.1 - 3 - 4 647 70
          AK293399.1 - 3 - 4 652 70
          AK297194.1 - 5 - 6 831 70
          AK312563.1 - 4 - 5 841 70
          AL832529.1 - 4 - 5 750 70
          BC022522.1 - 4 - 5 758 4794050 70
          BC031103.1 - 4 - 5 734 5301702 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300013859.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03000572.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CD200

          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          cell recognition
          negative regulation of macrophage activation
          regulation of immune response
          receptor activity
          receptor binding
          protein binding
          protein homodimerization activity
          cell adhesion molecule binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03010390.hg.1 TC03000572.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03010390.hg.1 ENST00000315711.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03010390.hg.1 ENST00000383681.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03010390.hg.1 ENST00000473539.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03010390.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03010390.hg.1 NM_005944.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000383681.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000383681.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000383681.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000383681.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000383681.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000383681.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000478595.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000478595.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000478595.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000478595.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000478595.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000478595.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000498096.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000498096.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000498096.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000498096.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000498096.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000498096.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000607516.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000607516.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000607516.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000607516.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000607516.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03010391.hg.1 TC03000572.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03010391.hg.1 ENST00000315711.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03010391.hg.1 ENST00000383681.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03010391.hg.1 ENST00000473539.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03010391.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03010391.hg.1 NM_005944.6 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000383681.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000383681.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000383681.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000383681.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000383681.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000383681.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000478595.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000478595.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000478595.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000478595.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000478595.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000478595.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000498096.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000498096.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000498096.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000498096.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000498096.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000498096.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000607516.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000607516.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000607516.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000607516.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000607516.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
          Affymetrix Human Clariom D Assay PSR0300205057.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205057.hg.1 ENST00000383681.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205057.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205057.hg.1 ENST00000478595.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205057.hg.1 ENST00000498096.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205057.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300205057.hg.1 NM_005944.6 Refseq
          Affymetrix Human Clariom D Assay PSR0300205057.hg.1 ENST00000315711.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205057.hg.1 ENST00000383681.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205057.hg.1 ENST00000473539.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205057.hg.1 ENST00000478595.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205057.hg.1 ENST00000498096.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205057.hg.1 NM_001004196.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300205057.hg.1 NM_005944.6 Refseq
          Affymetrix Human Clariom D Assay PSR0300205057.hg.1 TC0300013859.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline