| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21006146.hg.1 |
ENST00000291560.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21006146.hg.1 |
ENST00000542962.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21006146.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21006146.hg.1 |
TC21000508.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2100156374.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156374.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156374.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156374.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156374.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156374.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156374.hg.1 |
TC2100008306.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000291560.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
ENST00000443485.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008306.hg.1 |
NM_007031.1 |
Refseq |