Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01037093_m1
          See other UNG GE Assays ›
          Gene Symbol:
          UNG
          Gene Name:
          uracil DNA glycosylase
          Gene Aliases:
          DGU, HIGM4, HIGM5, UDG, UNG1, UNG15, UNG2
          Chromosome Location:
          Chr.12: 109097597 - 109110992 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01037093_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: UNG
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003362.4 NP_003353.1 5 - 6 932 60
          NM_080911.3 NP_550433.1 6 - 7 887 60
          GenBank mRNA AK291341.1 - 5 - 6 910 60
          AK299416.1 - 6 - 7 830 60
          AK313552.1 - 6 - 7 887 60
          BC015205.1 - 5 - 6 869 60
          BC050634.2 - 5 - 6 880 60
          CR541673.1 - 5 - 6 60
          CR749342.1 - 4 - 5 856 60
          X15653.1 - 5 - 6 883 60
          Y09008.1 - 6 - 7 874 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200008792.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000843.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene UNG

          Gene Ontology Categories:

          Function(s) Process(es)

          single strand break repair
          DNA repair
          base-excision repair
          cellular response to DNA damage stimulus
          somatic hypermutation of immunoglobulin genes
          somatic recombination of immunoglobulin gene segments
          negative regulation of apoptotic process
          depyrimidination
          isotype switching
          base-excision repair, AP site formation via deaminated base removal
          damaged DNA binding
          uracil DNA N-glycosylase activity
          protein binding
          hydrolase activity
          hydrolase activity, hydrolyzing N-glycosyl compounds
          ribosomal small subunit binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000540158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000540158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000540158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 ENST00000540158.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12010812.hg.1 ENST00000242576.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12010812.hg.1 ENST00000336865.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12010812.hg.1 ENST00000537518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12010812.hg.1 NM_003362.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12010812.hg.1 NM_080911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12010812.hg.1 TC12000843.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12010813.hg.1 ENST00000242576.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12010813.hg.1 ENST00000336865.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12010813.hg.1 NM_003362.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12010813.hg.1 NM_080911.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12010813.hg.1 TC12000843.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200164970.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164970.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164970.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164970.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164970.hg.1 NM_003362.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200164970.hg.1 NM_080911.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200164970.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164970.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164970.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164970.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164970.hg.1 NM_003362.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200164970.hg.1 NM_080911.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200164970.hg.1 TC1200008792.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000540158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000540158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000540158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 ENST00000540158.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_003362.3 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008792.hg.1 NM_080911.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200164969.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164969.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164969.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164969.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164969.hg.1 NM_003362.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200164969.hg.1 NM_080911.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200164969.hg.1 ENST00000242576.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164969.hg.1 ENST00000336865.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164969.hg.1 ENST00000446767.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164969.hg.1 ENST00000539287.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200164969.hg.1 NM_003362.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200164969.hg.1 NM_080911.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200164969.hg.1 TC1200008792.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0