| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000473460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000473460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000473460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000473460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000481735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000481735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000481735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000481735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000484893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000484893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000484893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000484893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000484893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
ENST00000484893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002130.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002130.hg.1 |
ENST00000429262.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002130.hg.1 |
ENST00000484893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002130.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002130.hg.1 |
TC20000169.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2000147891.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000147891.hg.1 |
ENST00000473460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000147891.hg.1 |
ENST00000484893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000147891.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000147891.hg.1 |
TC2000007016.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000147891.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000147891.hg.1 |
ENST00000473460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000147891.hg.1 |
ENST00000484893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000147891.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000262460.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000473460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000473460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000473460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000473460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000481735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000481735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000481735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000481735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000484893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000484893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000484893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000484893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000484893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
ENST00000484893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007016.hg.1 |
NM_021067.4 |
Refseq |