| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007271.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007271.hg.1 |
ENST00000372165.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007271.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007271.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06007271.hg.1 |
TC06000607.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008104.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156631.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156631.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156631.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156631.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156631.hg.1 |
ENST00000372163.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156631.hg.1 |
ENST00000372165.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600156631.hg.1 |
NM_001193341.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156631.hg.1 |
NM_152732.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600156631.hg.1 |
TC0600008104.hg.1 |
Clariom D TC |