| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023735.hg.1 |
ENST00000293405.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023735.hg.1 |
ENST00000377000.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023735.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023735.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19023735.hg.1 |
TC19001743.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000594854.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000594854.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000594854.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000594854.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000594854.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000594854.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000594854.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000594854.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011190.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900193968.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900193968.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900193968.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900193968.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900193968.hg.1 |
ENST00000293405.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900193968.hg.1 |
ENST00000486050.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900193968.hg.1 |
ENST00000600293.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900193968.hg.1 |
NM_152358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900193968.hg.1 |
TC1900011190.hg.1 |
Clariom D TC |