| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000525452.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000525452.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000525452.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000525452.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000525452.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000529820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000529820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000529820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000529820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000547403.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000547403.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000547403.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000547403.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000547403.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000547403.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000547836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000547836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000547836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000547836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000547836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000547836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12023540.hg.1 |
ENST00000532498.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12023540.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12023540.hg.1 |
TC12001785.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000433494.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000525452.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000525452.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000525452.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000525452.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000525452.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000529820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000529820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000529820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000529820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000532628.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000547403.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000547403.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000547403.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000547403.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000547403.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000547403.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000547836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000547836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000547836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000547836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000547836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000547836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011423.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200188468.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200188468.hg.1 |
ENST00000547836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200188468.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200188468.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200188468.hg.1 |
ENST00000532498.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200188468.hg.1 |
ENST00000547836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200188468.hg.1 |
ENST00000552392.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200188468.hg.1 |
NM_001100917.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200188468.hg.1 |
TC1200011423.hg.1 |
Clariom D TC |