Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01045843_m1
          See other VDR GE Assays ›
          Gene Symbol:
          VDR
          Gene Name:
          vitamin D receptor
          Gene Aliases:
          NR1I1, PPP1R163
          Chromosome Location:
          Chr.12: 47841537 - 47904994 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01045843_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: VDR
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000376.3 NP_000367.1 3 - 4 270 85
          NM_001017535.2 NP_001017535.1 4 - 5 392 85
          NM_001017536.2 NP_001017536.1 3 - 4 661 85
          NM_001364085.2 NP_001351014.1 3 - 4 270 85
          NM_001374661.1 NP_001361590.1 3 - 4 702 85
          NM_001374662.1 NP_001361591.1 2 - 3 189 85
          XM_024449178.2 XP_024304946.1 2 - 3 580 85
          XM_047429500.1 XP_047285456.1 5 - 6 469 85
          GenBank mRNA AB307700.1 - 1 - 2 147 85
          AF026260.1 - 1 - 2 147 85
          AK225844.1 - 3 - 4 306 85
          AK312267.1 - 3 - 4 299 85
          BC033465.1 - 3 - 4 286 85
          BC060832.1 - 3 - 4 670 85
          CF138099.1 - 3 - 4 297 85
          DA721726.1 - 4 - 5 438 85
          HQ692836.1 - 1 - 2 147 85
          J03258.1 - 3 - 4 262 85
          M65208.1 - 1 - 2 78 85
          X67482.1 - 2 - 3 197 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200010559.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001439.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene VDR

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell morphogenesis
          skeletal system development
          regulation of transcription, DNA-templated
          calcium ion transport
          cellular calcium ion homeostasis
          lactation
          negative regulation of cell proliferation
          organ morphogenesis
          positive regulation of gene expression
          negative regulation of keratinocyte proliferation
          cell differentiation
          positive regulation of bone mineralization
          intracellular steroid hormone receptor signaling pathway
          intracellular receptor signaling pathway
          phosphate ion transmembrane transport
          intracellular bile acid receptor signaling pathway
          mRNA transcription from RNA polymerase II promoter
          cell-cell junction maintenance
          positive regulation of keratinocyte differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          decidualization
          retinoic acid receptor signaling pathway
          intestinal absorption
          apoptotic process involved in mammary gland involution
          positive regulation of apoptotic process involved in mammary gland involution
          mammary gland branching involved in pregnancy
          vitamin D receptor signaling pathway
          positive regulation of vitamin D receptor signaling pathway
          response to bile acid
          negative regulation of phosphate transmembrane transport
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          vitamin D binding
          protein binding
          zinc ion binding
          lithocholic acid receptor activity
          sequence-specific DNA binding
          metal ion binding
          retinoid X receptor binding
          vitamin D response element binding
          calcitriol binding
          lithocholic acid binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000548664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000548664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000548664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000548664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000548664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000548664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000548664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline