| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000288098.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000288098.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000288098.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000288098.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000288098.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000288098.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000288098.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000288098.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000288098.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000288098.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000429149.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000429149.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000429149.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000429149.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000429149.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000429149.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000429149.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000429149.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000429149.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000429149.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000429149.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000563721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000563721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000563721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000563721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000563721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000566361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000566361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000566361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000566361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000566361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000566361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000566361.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000569641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000569641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000569641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000569641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000574181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000574181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000574181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000574181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
ENST00000574181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172771.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_001172772.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_152456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_152456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_152456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_152456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_152456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_152456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_152456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_152456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_152456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_152456.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008278.hg.1 |
NM_152456.2 |
Refseq |