| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528635 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528735 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962250 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962245 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962170 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962265 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962217 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962174 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962239 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962219 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962196 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528648 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528721 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962227 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962235 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962173 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528743 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528636 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962184 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962201 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962229 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528711 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962211 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962261 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528704 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962263 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528730 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962246 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528659 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528728 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962238 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528645 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962188 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962218 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962273 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528664 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962209 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528712 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962208 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962197 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962251 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962262 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528683 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962187 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962205 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962271 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528651 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962189 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528726 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962228 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962266 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962274 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962195 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962222 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962225 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528713 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962215 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528731 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528639 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962268 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962175 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962192 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962247 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962232 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528754 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962207 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962269 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962234 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962236 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962171 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528732 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962223 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528747 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962180 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962190 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962214 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962258 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962199 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962226 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528663 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962212 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962203 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962183 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528692 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528688 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962210 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962275 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962200 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962177 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528656 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08528655 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P28962248 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
121518 |
AK023216.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
121518 |
AK126223.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
121518 |
BC004415.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
121518 |
BC006291.2 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
121518 |
BC007774.2 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
121518 |
BC021554.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
121518 |
CR622733.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
121518 |
NM_024598.2 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
31799 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 8x60K |
11311 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
558704 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
696492 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
121143 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
973009 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P368870 |
- |
- |
| Agilent Whole Human Genome Oligo Microarray 4x44K |
20020 |
- |
- |
| Agilent Human Genome CGH Microarray 44B |
A_14_P122953 |
- |
- |
| Agilent Human Genome CGH Microarray 44A |
A_14_P122953 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
18683 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_14_P122953 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P39767686 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P56899005 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P368870 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P122953 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_16_P39767686 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P122953 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P39767723 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P19634599 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P39767686 |
- |
- |
| Agilent Human Genome CGH Microarray 4x44K |
A_14_P122953 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
67283 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
51610 |
- |
- |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
90029 |
- |
- |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
15989 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P11552892 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P19634624 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P39767714 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P11552129 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P19634599 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P39767723 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P122953 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P19634592 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P19634615 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P39767686 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P19634600 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P122953 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P19634599 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P39767686 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P19634600 |
- |
- |