| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023166.hg.1 |
ENST00000284694.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023166.hg.1 |
ENST00000356858.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023166.hg.1 |
ENST00000454341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023166.hg.1 |
ENST00000544758.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023166.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023166.hg.1 |
TC10001747.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000463082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000463082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000463082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000463082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000463082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000488181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000488181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000488181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000488181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
ENST00000488181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000368674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000463082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000463082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000463082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000463082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000463082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000488181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000488181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000488181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000488181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
ENST00000488181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012198.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000183390.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000183390.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000183390.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000183390.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000183390.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000183390.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000183390.hg.1 |
ENST00000284694.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000183390.hg.1 |
ENST00000356858.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000183390.hg.1 |
ENST00000424927.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000183390.hg.1 |
ENST00000432642.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000183390.hg.1 |
ENST00000480379.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000183390.hg.1 |
NM_001004298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000183390.hg.1 |
TC1000012198.hg.1 |
Clariom D TC |