| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X001447.hg.1 |
ENST00000379341.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X001447.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X001447.hg.1 |
TC0X000110.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X001446.hg.1 |
ENST00000379341.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X001446.hg.1 |
ENST00000495599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X001446.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X001446.hg.1 |
TC0X000110.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00146500.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146500.hg.1 |
ENST00000379349.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146500.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146500.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00146500.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146500.hg.1 |
ENST00000379349.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146500.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146500.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00146500.hg.1 |
TC0X00006794.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00146499.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146499.hg.1 |
ENST00000379349.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146499.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146499.hg.1 |
ENST00000495599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146499.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00146499.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146499.hg.1 |
ENST00000379349.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146499.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146499.hg.1 |
ENST00000495599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00146499.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00146499.hg.1 |
TC0X00006794.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |