| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10004878.hg.1 |
ENST00000373290.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10004878.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10004878.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10004878.hg.1 |
TC10000420.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10004876.hg.1 |
ENST00000373290.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10004876.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10004876.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10004876.hg.1 |
TC10000420.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000470508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000470508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000470508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000470508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000470508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000470508.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000478112.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000478112.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000478112.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000478112.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000478112.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000153582.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153582.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153582.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153582.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153582.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153582.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153582.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000153582.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153582.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153582.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153582.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153582.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153582.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153582.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000153582.hg.1 |
TC1000007895.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000470508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000470508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000470508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000470508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000470508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000470508.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000478112.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000478112.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000478112.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000478112.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000478112.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007895.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000153581.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153581.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153581.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153581.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153581.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153581.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153581.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000153581.hg.1 |
ENST00000373290.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153581.hg.1 |
ENST00000452130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153581.hg.1 |
ENST00000459981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153581.hg.1 |
ENST00000475069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153581.hg.1 |
ENST00000486093.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153581.hg.1 |
ENST00000490083.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153581.hg.1 |
NM_012339.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000153581.hg.1 |
TC1000007895.hg.1 |
Clariom D TC |