Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01057139_g1
          See other FTO GE Assays ›
          Gene Symbol:
          FTO
          Gene Name:
          FTO alpha-ketoglutarate dependent dioxygenase
          Gene Aliases:
          ALKBH9, BMIQ14, GDFD, IFEX9
          Chromosome Location:
          Chr.16: 53703963 - 54121941 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01057139_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FTO
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080432.3 NP_001073901.1 2 - 3 154 108
          NM_001363891.2 NP_001350820.1 2 - 3 347 108
          NM_001363894.2 NP_001350823.1 2 - 3 347 108
          NM_001363896.2 NP_001350825.1 2 - 3 347 108
          NM_001363898.2 NP_001350827.1 2 - 3 347 108
          NM_001363899.2 NP_001350828.1 2 - 3 347 108
          NM_001363900.2 NP_001350829.1 2 - 3 347 108
          NM_001363901.2 NP_001350830.1 2 - 3 347 108
          NM_001363903.2 NP_001350832.1 2 - 3 347 108
          NM_001363988.2 NP_001350917.1 2 - 3 347 108
          NM_001438128.1 2 - 3 154 108
          NM_001438129.1 2 - 3 154 108
          NM_001438130.1 2 - 3 154 108
          XM_011523314.4 XP_011521616.1 2 - 3 347 108
          XM_011523315.4 XP_011521617.1 2 - 3 347 108
          XM_011523316.4 XP_011521618.1 2 - 3 347 108
          XM_017023654.3 XP_016879143.1 2 - 3 347 108
          XM_017023656.3 XP_016879145.1 2 - 3 347 108
          XM_017023657.3 XP_016879146.1 2 - 3 347 108
          XM_024450437.2 XP_024306205.1 2 - 3 347 108
          XM_047434606.1 XP_047290562.1 2 - 3 347 108
          XM_047434607.1 XP_047290563.1 2 - 3 347 108
          XM_047434608.1 XP_047290564.1 2 - 3 347 108
          XM_047434609.1 XP_047290565.1 2 - 3 347 108
          GenBank mRNA AB051539.1 - 2 - 3 131 108
          AK096554.1 - 2 - 3 154 108
          DA324897.1 - 2 - 3 348 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600007893.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16000449.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          DNA dealkylation involved in DNA repair
          regulation of lipid storage
          snRNA processing
          regulation of multicellular organism growth
          RNA repair
          regulation of respiratory system process
          adipose tissue development
          mRNA destabilization
          regulation of white fat cell proliferation
          regulation of brown fat cell differentiation
          protein binding
          ferrous iron binding
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
          transferase activity
          oxidative RNA demethylase activity
          oxidative DNA demethylase activity
          metal ion binding
          dioxygenase activity
          RNA N6-methyladenosine dioxygenase activity
          tRNA demethylase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000268349.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000268349.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000268349.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000268349.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000268349.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000268349.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000268349.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000268349.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000431610.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000431610.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000431610.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000431610.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000431610.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000431610.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000431610.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000460382.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000460382.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000460382.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000460382.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000460382.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000460382.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000460382.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000460382.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000463855.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000463855.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000463855.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000463855.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000463855.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000463855.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000463855.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000463855.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000463855.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000463855.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000464071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000464071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000464071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000464071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000464071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000464071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000464071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000464071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000464071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000464071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000471389.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000472835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000472835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000472835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000472835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000472835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000570395.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000570395.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000570395.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000570395.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000570395.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 ENST00000570395.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600007893.hg.1 NM_001080432.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012384.hg.1 ENST00000419363.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012384.hg.1 ENST00000419363.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012384.hg.1 ENST00000419363.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012384.hg.1 ENST00000419363.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012384.hg.1 ENST00000419363.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012384.hg.1 ENST00000419363.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline