| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000586874.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000586874.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000586874.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000586874.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000586874.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000586874.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589003.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589003.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589003.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589003.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589003.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589003.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000593249.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000593249.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000593249.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000593249.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
ENST00000593249.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009728.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009728.hg.1 |
ENST00000393853.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009728.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009728.hg.1 |
TC17000745.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000586874.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000586874.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000586874.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000586874.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000586874.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000586874.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589003.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589003.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589003.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589003.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589003.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589003.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000593249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000593249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000593249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000593249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
ENST00000593249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008514.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
ENST00000586874.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
ENST00000589003.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
ENST00000593249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
ENST00000240335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
ENST00000393853.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
ENST00000586874.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
ENST00000589003.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
ENST00000589449.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
ENST00000590174.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
ENST00000593249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
NM_018488.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700163525.hg.1 |
TC1700008514.hg.1 |
Clariom D TC |