Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01057837_m1
          See other HAPSTR1 GE Assays ›
          Gene Symbol:
          HAPSTR1
          Gene Name:
          HUWE1 associated protein modifying stress responses
          Gene Aliases:
          C16orf72, PRO0149, TAPR1
          Chromosome Location:
          Chr.16: 9091644 - 9121635 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01057837_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HAPSTR1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014117.3 NP_054836.2 2 - 3 714 82
          XM_011522462.4 XP_011520764.1 2 - 3 714 82
          GenBank mRNA BC113570.1 - 2 - 3 578 82
          CB959956.1 - 2 - 3 498 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600006842.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16000150.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HAPSTR1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cellular response to stress
          negative regulation of signal transduction by p53 class mediator
          protein binding
          ubiquitin protein ligase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR16002338.hg.1 ENST00000327827.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16002338.hg.1 NM_014117.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16002338.hg.1 TC16000150.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 ENST00000574616.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006842.hg.1 NM_014117.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600148763.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Clariom D Assay PSR1600148763.hg.1 ENST00000574285.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600148763.hg.1 NM_014117.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600148763.hg.1 ENST00000327827.11 Ensembl
          Affymetrix Human Clariom D Assay PSR1600148763.hg.1 ENST00000574285.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1600148763.hg.1 NM_014117.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600148763.hg.1 TC1600006842.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline