| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009014.hg.1 |
ENST00000280325.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009014.hg.1 |
ENST00000280325.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009014.hg.1 |
ENST00000280325.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009014.hg.1 |
ENST00000280325.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009014.hg.1 |
ENST00000280325.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009014.hg.1 |
ENST00000280325.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009014.hg.1 |
NM_198498.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009014.hg.1 |
NM_198498.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009014.hg.1 |
NM_198498.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009014.hg.1 |
NM_198498.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009014.hg.1 |
NM_198498.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009014.hg.1 |
NM_198498.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012330.hg.1 |
ENST00000280325.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012330.hg.1 |
NM_198498.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012330.hg.1 |
TC11000989.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1100009014.hg.1 |
ENST00000280325.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009014.hg.1 |
ENST00000280325.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009014.hg.1 |
ENST00000280325.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009014.hg.1 |
ENST00000280325.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009014.hg.1 |
ENST00000280325.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009014.hg.1 |
ENST00000280325.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009014.hg.1 |
NM_198498.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009014.hg.1 |
NM_198498.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009014.hg.1 |
NM_198498.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009014.hg.1 |
NM_198498.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009014.hg.1 |
NM_198498.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009014.hg.1 |
NM_198498.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100166626.hg.1 |
ENST00000280325.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100166626.hg.1 |
NM_198498.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100166626.hg.1 |
ENST00000280325.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100166626.hg.1 |
NM_198498.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100166626.hg.1 |
TC1100009014.hg.1 |
Clariom D TC |