Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01057884_s1
          See other TMEM203 GE Assays ›
          Gene Symbol:
          TMEM203
          Gene Name:
          transmembrane protein 203
          Gene Aliases:
          HBEBP1
          Chromosome Location:
          Chr.9: 137204082 - 137205638 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01057884_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TMEM203
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053045.1 NP_444273.1 1 1490 70
          GenBank mRNA AX775751.1 - 1 1499 70
          BC009283.2 - 1 1490 4298098 70
          BC009461.1 - 1 1490 3538818 70
          BC067786.1 - 1 1502 5275460 70

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TMEM203

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          cellular calcium ion homeostasis
          spermatogenesis
          mitophagy in response to mitochondrial depolarization
          xenophagy
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 95301 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3285024 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P136238 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P136238 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 7748 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 2861 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 30913 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3285024 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P136238 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3285024 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P136238 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P136238 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P136238 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012076 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012075 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16279288 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012068 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012060 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16279290 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012078 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16279283 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16279284 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16279292 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012065 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012050 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012073 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012057 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16279281 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012070 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012077 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16279294 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012059 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012048 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012067 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012055 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07012053 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_053045_at - -
          Affymetrix Human Gene 1.0 ST Array 8165487 - -
          Affymetrix Human Gene 1.0 ST Array 8165489 - -
          Affymetrix Human Gene 1.0 ST Array 8165491 - -
          Affymetrix Human Gene 1.0 ST Array 8165643 - -
          Affymetrix Human Gene 1.0 ST Array 8165488 - -
          Affymetrix Human Gene 1.0 ST Array 8165490 - -
          Affymetrix Human Gene 1.1 ST Array 8165642 - -
          Affymetrix Human Gene 1.1 ST Array 8165486 - -
          Affymetrix HT HG-U133+ PM Array Plate 225052_PM_at - -
          Affymetrix Human Genome U219 Array 11753774_a_at - -
          Affymetrix Human Genome U219 Array 11753774_PM_a_at - -
          Human Genome U133 Plus 2.0 Array 225052_at - -
          Human Genome U133 Plus 2.0 Array 225052_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225052_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225052_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225052_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225052_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225052_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225052_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225052_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225052_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225052_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225052_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225052_at1 - -
          Human Genome U95 Set 45759_PM_g_at - -
          Human Genome U95 Set 45758_PM_at - -
          Human Genome U95 Set 45759_g_at - -
          Human Genome U95 Set 45758_at - -
          Human Genome U133 Set 225052_at - -
          Human Genome U133 Set 225052_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038262 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038262 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 134886 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2073010 AX775751.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2073010 BC009283.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2073010 BC009461.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2073010 BC067786.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2073010 NM_053045.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2073012 AX775751.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2073012 BC009283.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2073012 BC009461.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2073012 BC067786.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2073012 NM_053045.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1672822 - -
          HumanHT-12 V4.0 expression beadchip ILMN_2073010 AX775751.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2073010 BC009283.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2073010 BC009461.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2073010 BC067786.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2073010 NM_053045.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2073012 AX775751.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2073012 BC009283.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2073012 BC009461.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2073012 BC067786.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2073012 NM_053045.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2073012 AX775751.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2073012 BC009283.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2073012 BC009461.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2073012 BC067786.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2073012 NM_053045.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2073012 AX775751.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2073012 BC009283.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2073012 BC009461.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2073012 BC067786.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2073012 NM_053045.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_16596685-S AX775751.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_16596685-S BC009283.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_16596685-S BC009461.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_16596685-S BC067786.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_16596685-S NM_053045.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_864-S - -
          Illumina human-6 v1.0 expression beadchip 0001690500 - -
          Illumina humanRef-8 v1.0 expression beadchip 0001690500 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1672868 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1672822 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1672822 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 6590075 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_864 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2073012 AX775751.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2073012 BC009283.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2073012 BC009461.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2073012 BC067786.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2073012 NM_053045.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2073010 AX775751.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2073010 BC009283.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2073010 BC009461.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2073010 BC067786.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2073010 NM_053045.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2073012 AX775751.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2073012 BC009283.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2073012 BC009461.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2073012 BC067786.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2073012 NM_053045.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2073010 AX775751.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2073010 BC009283.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2073010 BC009461.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2073010 BC067786.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2073010 NM_053045.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2073012 AX775751.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2073012 BC009283.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2073012 BC009461.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2073012 BC067786.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2073012 NM_053045.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1672868 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1672822 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2073012 AX775751.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2073012 BC009283.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2073012 BC009461.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2073012 BC067786.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2073012 NM_053045.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline