Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01058307_m1
          See other ACMSD GE Assays ›
          Gene Symbol:
          ACMSD
          Gene Name:
          aminocarboxymuconate semialdehyde decarboxylase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 134838616 - 134902034 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01058307_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ACMSD
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001307983.2 NP_001294912.1 5 - 6 612 62
          NM_138326.3 NP_612199.2 5 - 6 555 62
          XM_005263586.5 XP_005263643.1 3 - 4 408 62
          XM_005263588.5 XP_005263645.1 4 - 5 505 62
          XM_005263590.5 XP_005263647.1 2 - 3 331 62
          XM_011510592.3 XP_011508894.1 5 - 6 618 62
          XM_017003325.2 XP_016858814.1 5 - 6 615 62
          XM_017003326.2 XP_016858815.1 5 - 6 2626 62
          XM_047443333.1 XP_047299289.1 4 - 5 521 62
          GenBank mRNA AB071418.1 - 5 - 6 488 62
          AK125008.1 - 5 - 6 555 62
          BC016018.1 - 5 - 6 614 62
          BC107420.1 - 5 - 6 495 62
          KU178859.1 - 2 - 3 314 62
          KU178860.1 - 2 - 3 314 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200009424.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000877.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ACMSD

          Gene Ontology Categories:

          Function(s) Process(es)

          tryptophan catabolic process
          secondary metabolic process
          negative regulation of quinolinate biosynthetic process
          picolinic acid biosynthetic process
          regulation of 'de novo' NAD biosynthetic process from tryptophan
          aminocarboxymuconate-semialdehyde decarboxylase activity
          protein binding
          zinc ion binding
          hydrolase activity
          lyase activity
          carboxy-lyase activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000498093.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000498093.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000498093.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000498093.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02012943.hg.1 ENST00000283054.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02012943.hg.1 ENST00000356140.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02012943.hg.1 ENST00000392928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02012943.hg.1 NM_138326.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02012943.hg.1 TC02000877.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000498093.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000498093.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000498093.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000498093.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200165889.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200165889.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200165889.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200165889.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200165889.hg.1 NM_138326.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200165889.hg.1 ENST00000356140.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200165889.hg.1 ENST00000392928.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200165889.hg.1 ENST00000485893.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200165889.hg.1 NM_001307983.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200165889.hg.1 NM_138326.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200165889.hg.1 TC0200009424.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline