| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000424938.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000424938.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000424938.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000431363.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000431363.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000431363.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000431363.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000542716.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000542716.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
ENST00000542716.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
NR_028594.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
NR_028594.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
NR_028594.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
NR_028594.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007043.hg.1 |
NR_028594.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016774.hg.1 |
ENST00000397732.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016774.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016774.hg.1 |
TC19002684.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016775.hg.1 |
ENST00000397732.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016775.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016775.hg.1 |
TC19002684.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016772.hg.1 |
ENST00000397732.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016772.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016772.hg.1 |
TC19002684.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016776.hg.1 |
ENST00000397732.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016776.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016776.hg.1 |
TC19002684.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
ENST00000455490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
ENST00000455490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
ENST00000455490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
ENST00000455490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
ENST00000493776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
ENST00000493776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016773.hg.1 |
ENST00000397732.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016773.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016773.hg.1 |
TC19002684.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016777.hg.1 |
ENST00000397732.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016777.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016777.hg.1 |
TC19002684.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
ENST00000455490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
ENST00000455490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
ENST00000455490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
ENST00000455490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
ENST00000493776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
ENST00000493776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011885.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204563.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204563.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204563.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204563.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204563.hg.1 |
TC1900011885.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900204568.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204568.hg.1 |
ENST00000455490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204568.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204568.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204568.hg.1 |
ENST00000455490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204568.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204568.hg.1 |
TC1900011885.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900204561.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204561.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204561.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204561.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204561.hg.1 |
TC1900011885.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900204570.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204570.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204570.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204570.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204570.hg.1 |
TC1900011885.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900204562.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204562.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204562.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204562.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204562.hg.1 |
TC1900011885.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900204564.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204564.hg.1 |
ENST00000455490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204564.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204564.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204564.hg.1 |
ENST00000455490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204564.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204564.hg.1 |
TC1900011885.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900204566.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204566.hg.1 |
ENST00000455490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204566.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204566.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204566.hg.1 |
ENST00000455490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204566.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204566.hg.1 |
TC1900011885.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000344893.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000424938.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000424938.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000424938.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000431363.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000431363.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000431363.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000431363.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000542716.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000542716.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
ENST00000542716.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
NR_028594.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
NR_028594.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
NR_028594.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
NR_028594.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007043.hg.1 |
NR_028594.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204571.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204571.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204571.hg.1 |
ENST00000397732.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204571.hg.1 |
NM_152601.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204571.hg.1 |
TC1900011885.hg.1 |
Clariom D TC |