| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003966.hg.1 |
ENST00000303775.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003966.hg.1 |
ENST00000351429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003966.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003966.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003966.hg.1 |
TC21000350.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007846.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100152888.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100152888.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100152888.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100152888.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100152888.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100152888.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100152888.hg.1 |
ENST00000303775.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100152888.hg.1 |
ENST00000351429.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100152888.hg.1 |
ENST00000460212.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100152888.hg.1 |
NM_013240.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100152888.hg.1 |
NM_182749.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100152888.hg.1 |
NR_047510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100152888.hg.1 |
TC2100007846.hg.1 |
Clariom D TC |