| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11013293.hg.1 |
ENST00000361763.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11013293.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11013293.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11013293.hg.1 |
TC11001060.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000528435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000528435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000528435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000528435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000528435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000528435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000529713.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000529713.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000529713.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000529713.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000529713.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100168378.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168378.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168378.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168378.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168378.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100168378.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168378.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168378.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168378.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168378.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100168378.hg.1 |
TC1100009200.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100168377.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168377.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168377.hg.1 |
ENST00000528435.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168377.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168377.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168377.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100168377.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168377.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168377.hg.1 |
ENST00000528435.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168377.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168377.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100168377.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100168377.hg.1 |
TC1100009200.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000361763.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000526146.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000528435.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000528435.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000528435.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000528435.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000528435.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000528435.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000528600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000529713.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000529713.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000529713.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000529713.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000529713.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
ENST00000531913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009200.hg.1 |
NM_000733.3 |
Refseq |