Search
Search
| Gene Symbol: IRF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195286.1 | NP_001182215.1 | 4 - 5 | 618 | 99 | |
| NR_046000.2 | 4 - 5 | 618 | 99 | |||
| GenBank mRNA | D78261.1 | - | 4 - 5 | 587 | 99 | |
| U52682.1 | - | 4 - 5 | 617 | 99 | ||
Set Membership: |
|