Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01063426_s1
          See other SMIM10 GE Assays ›
          Gene Symbol:
          SMIM10
          Gene Name:
          small integral membrane protein 10
          Gene Aliases:
          CXorf69
          Chromosome Location:
          Chr.X: 134990938 - 134992473 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01063426_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SMIM10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163438.1 NP_001156910.1 1 1271 97
          GenBank mRNA BQ016562.1 - 1 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00008492.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000644.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32702927 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11971068 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17399922 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32702950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32702947 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32702941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11971060 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11971056 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32702929 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32702925 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32702945 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11971059 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32702930 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32702943 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32702949 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32702939 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32702948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11971062 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32702933 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32702944 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P67534 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 7601 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 22527 - -
          Agilent Human Genome CGH Microarray 44B A_14_P120465 - -
          Agilent Human Genome CGH Microarray 44A A_14_P120465 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 37786 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P67534 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P67534 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702958 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17399922 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702944 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702930 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702960 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702939 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971074 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702949 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971082 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971073 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702957 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702929 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971080 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702943 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702945 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971068 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702941 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971062 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702930 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971074 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702944 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702960 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702943 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702958 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971062 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702957 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971068 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971080 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702939 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971073 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17399922 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971082 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702941 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702929 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702945 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11971060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32702949 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P120465 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P120465 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P120465 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 81469 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 81247 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P120465 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 8170007 - -
          Affymetrix HT HG-U133+ PM Array Plate 227976_PM_at - -
          Affymetrix Human Genome U219 Array 11754398_at - -
          Affymetrix Human Genome U219 Array 11754397_PM_at - -
          Affymetrix Human Genome U219 Array 11754398_PM_at - -
          Affymetrix Human Genome U219 Array 11754397_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0X00008492.hg.1 ENST00000330288.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008492.hg.1 ENST00000330288.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008492.hg.1 ENST00000330288.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008492.hg.1 NM_001163438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008492.hg.1 NM_001163438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008492.hg.1 NM_001163438.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X008437.hg.1 TC0X000644.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0X008437.hg.1 ENST00000330288.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X008437.hg.1 NM_001163438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00157697.hg.1 ENST00000330288.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157697.hg.1 NM_001163438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00157697.hg.1 ENST00000330288.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157697.hg.1 NM_001163438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00157697.hg.1 TC0X00008492.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00157695.hg.1 ENST00000330288.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157695.hg.1 NM_001163438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00157695.hg.1 ENST00000330288.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157695.hg.1 NM_001163438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00157695.hg.1 TC0X00008492.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00157694.hg.1 ENST00000330288.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157694.hg.1 NM_001163438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00157694.hg.1 ENST00000330288.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157694.hg.1 NM_001163438.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00157694.hg.1 TC0X00008492.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0X00008492.hg.1 ENST00000330288.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008492.hg.1 ENST00000330288.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008492.hg.1 ENST00000330288.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008492.hg.1 NM_001163438.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008492.hg.1 NM_001163438.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008492.hg.1 NM_001163438.1 Refseq
          Human Genome U133 Plus 2.0 Array 227976_at - -
          Human Genome U133 Plus 2.0 Array 227976_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227976_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227976_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227976_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227976_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227976_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227976_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227976_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227976_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227976_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227976_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227976_at11 - -
          Human Genome U95 Set 45194_PM_at - -
          Human Genome U95 Set 45194_at - -
          Human Genome U95 Set 76329_at - -
          Human Genome U95 Set 76329_PM_at - -
          Human Genome U133 Set 227976_PM_at - -
          Human Genome U133 Set 227976_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A041566 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A041566 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 229361 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1683828 BC008642.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1683828 NM_001163438.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1683828 BC008642.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1683828 NM_001163438.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1683828 BC008642.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1683828 NM_001163438.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1683828 BC008642.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1683828 NM_001163438.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline