Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01063606_m1
          See other B9D1 GE Assays ›
          Gene Symbol:
          B9D1
          Gene Name:
          B9 domain containing 1
          Gene Aliases:
          B9, EPPB9, MKS9, MKSR1
          Chromosome Location:
          Chr.17: 19335372 - 19378193 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01063606_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B9D1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001243473.2 NP_001230402.1 3 - 4 321 67
          NM_001243475.2 NP_001230404.1 3 - 4 517 67
          NM_001321214.1 NP_001308143.1 3 - 4 414 67
          NM_001321215.1 NP_001308144.1 3 - 4 414 67
          NM_001321216.1 NP_001308145.1 3 - 4 414 67
          NM_001321217.1 NP_001308146.1 3 - 4 414 67
          NM_001321218.1 NP_001308147.1 3 - 4 414 67
          NM_001321219.1 NP_001308148.1 3 - 4 414 67
          NM_015681.4 NP_056496.1 3 - 4 414 67
          XM_005256607.2 XP_005256664.1 3 - 4 386 67
          XM_005256610.1 XP_005256667.1 3 - 4 389 67
          XM_017024452.1 XP_016879941.1 6 - 7 2778 67
          GenBank mRNA AB030506.1 - 3 - 4 403 67
          AK293815.1 - 3 - 4 387 67
          AK297782.1 - 3 - 4 304 67
          AK309320.1 - 3 - 4 506 67
          BC002944.2 - 3 - 4 345 3953287 67
          BI833202.1 - 3 - 4 409 67
          HY047032.1 - 2 - 3 345 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700010015.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001240.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene B9D1

          Panther Classification:

          Molecular Function -

          structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          vasculature development
          smoothened signaling pathway
          regulation of protein localization
          cilium assembly
          embryonic digit morphogenesis
          camera-type eye development
          cilium morphogenesis
          neuroepithelial cell differentiation
          hedgehog receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000261499.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000261499.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000261499.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000261499.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000261499.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000261499.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000261499.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000261499.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000261499.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000261499.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000261499.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000268841.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000268841.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000268841.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000268841.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000268841.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000268841.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000268841.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000268841.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395615.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395615.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395615.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395615.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395615.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395615.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395615.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395615.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395615.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395615.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395616.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395616.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395616.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395616.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395616.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395616.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395616.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395616.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395616.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395616.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000395616.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000440841.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000440841.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000440841.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000440841.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000440841.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000440841.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000461069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000461069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000461069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000461069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000461069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000461069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000461069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000461069.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000468679.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000468679.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000468679.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000468679.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000476298.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000476298.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000476298.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000476298.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000476298.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477478.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477478.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477478.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477478.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477478.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477478.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477478.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477478.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477478.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477478.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477478.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477478.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477478.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477683.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477683.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477683.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477683.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477683.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477683.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477683.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477683.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000477683.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000487415.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000487415.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000487415.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000487415.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000574508.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000574508.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000574508.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000574508.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000574508.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575403.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575403.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575403.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575403.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575403.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575403.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575403.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575403.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575403.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575403.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575478.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575478.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575478.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575478.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575478.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575478.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000575478.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000582857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000582857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000582857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000582857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000582857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000582857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 ENST00000582857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243475.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243475.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243475.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243475.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243475.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243475.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243475.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243475.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243475.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_001243475.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_015681.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_015681.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_015681.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_015681.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_015681.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_015681.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_015681.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_015681.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_015681.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_015681.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010015.hg.1 NM_015681.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline