Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01066102_g1
          See other TRPM2 GE Assays ›
          Gene Symbol:
          TRPM2
          Gene Name:
          transient receptor potential cation channel subfamily M member 2
          Gene Aliases:
          EREG1, KNP3, LTRPC2, LTrpC-2, NUDT9H, NUDT9L1, TRPC7
          Chromosome Location:
          Chr.21: 44350141 - 44442644 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01066102_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRPM2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001320350.2 NP_001307279.2 27 - 28 4104 86
          NM_001433516.1 NP_001420445.1 27 - 28 4109 86
          NM_003307.4 NP_003298.2 26 - 27 3954 86
          NR_038257.2 26 - 27 3957 86
          XM_011529736.3 XP_011528038.1 27 - 28 4044 86
          GenBank mRNA AB001535.1 - 26 - 27 4319 86
          AB166745.1 - 27 - 28 4087 86
          AB166746.1 - 22 - 23 3326 86
          AJ878416.1 - 26 - 27 3937 86
          AK301451.1 - 7 - 8 927 86
          BC112342.1 - 27 - 28 4037 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100007359.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000225.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TRPM2

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          dendritic cell chemotaxis
          ion transport
          sodium ion transport
          calcium ion transport
          manganese ion transport
          response to heat
          response to purine-containing compound
          calcium-mediated signaling
          regulation of actin cytoskeleton organization
          response to hydroperoxide
          ion transmembrane transport
          sodium ion transmembrane transport
          release of sequestered calcium ion into cytosol
          protein homotetramerization
          regulation of filopodium assembly
          transmembrane transport
          cellular response to hydrogen peroxide
          calcium ion transmembrane transport
          cellular response to calcium ion
          manganese ion transmembrane transport
          cellular response to temperature stimulus
          zinc II ion transmembrane transport
          dendritic cell differentiation
          calcium ion transmembrane import into cytosol
          calcium ion import across plasma membrane
          ion channel activity
          cation channel activity
          calcium channel activity
          sodium channel activity
          manganese ion transmembrane transporter activity
          calcium ion binding
          calcium-release channel activity
          hydrolase activity
          metal ion binding
          metal ion transmembrane transporter activity
          ADP-ribose diphosphatase activity
          mono-ADP-D-ribose binding
          ligand-gated calcium channel activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR21002703.hg.1 ENST00000300482.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21002703.hg.1 ENST00000397928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21002703.hg.1 ENST00000397932.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21002703.hg.1 ENST00000498430.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21002703.hg.1 NM_003307.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21002703.hg.1 NR_038257.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21002703.hg.1 TC21000225.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21002702.hg.1 ENST00000300481.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21002702.hg.1 ENST00000300482.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21002702.hg.1 ENST00000397928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21002702.hg.1 ENST00000397932.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21002702.hg.1 ENST00000498430.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21002702.hg.1 ENST00000540347.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21002702.hg.1 NM_003307.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21002702.hg.1 NR_038257.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21002702.hg.1 TC21000225.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2100149806.hg.1 ENST00000300482.9 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149806.hg.1 ENST00000397928.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149806.hg.1 ENST00000397932.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149806.hg.1 ENST00000498430.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149806.hg.1 NM_003307.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100149806.hg.1 NR_038257.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100149806.hg.1 ENST00000300482.9 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149806.hg.1 ENST00000397928.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149806.hg.1 ENST00000397932.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149806.hg.1 ENST00000498430.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149806.hg.1 NM_003307.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100149806.hg.1 NR_038257.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100149806.hg.1 TC2100007359.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 ENST00000300481.13 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 ENST00000300482.9 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 ENST00000397928.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 ENST00000397932.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 ENST00000490982.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 ENST00000498430.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 ENST00000621064.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 NM_003307.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 NR_038257.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 ENST00000300481.13 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 ENST00000300482.9 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 ENST00000397928.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 ENST00000397932.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 ENST00000490982.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 ENST00000498430.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 ENST00000621064.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 NM_003307.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 NR_038257.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100149805.hg.1 TC2100007359.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline