| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007202.hg.1 |
ENST00000207457.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007202.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007202.hg.1 |
TC01000466.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100154882.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100154882.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100154882.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100154882.hg.1 |
ENST00000207457.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100154882.hg.1 |
ENST00000469024.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100154882.hg.1 |
NM_014466.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100154882.hg.1 |
TC0100007790.hg.1 |
Clariom D TC |